Chemical Identifiers
CAS633-96-5
IUPAC Namesodium 4-[2-(2-oxo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]benzene-1-sulfonate
Molecular FormulaC16H11N2NaO4S
InChI KeyIGQFKZCLTLAWLO-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=C(NN=C2C(=O)C=CC3=CC=CC=C23)C=C1
View more