Chemical Identifiers
CAS1400-62-0
IUPAC Name4,12-bis[(2,4-dihydroxy-6-methylphenyl)amino]-3,13-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1,3,6,9,12-pentaene-5,11-dione
Molecular FormulaC28H24N2O7
InChI KeyVPEASJIRGSVXBF-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC(O)=C1NC1=C(C)C2=C3C(OC2=CC1=O)=CC(=O)C(NC1=C(C)C=C(O)C=C1O)=C3C
View more