Oxalacetic acid, 98%
Oxalacetic acid, 98%
Oxalacetic acid, 98%
Oxalacetic acid, 98%
Thermo Scientific Chemicals

Oxalacetic acid, 98%

CAS: 328-42-7 | C4H4O5 | 132.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 416600050
Price (USD)
67.65
Special offer
Online exclusive
Ends: 26-Sep-2025
83.70
Save 16.05 (19%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 26-Sep-2025
83.70
Save 16.05 (19%)
Each
Add to cart
Chemical Identifiers
CAS328-42-7
IUPAC Name2-oxobutanedioic acid
Molecular FormulaC4H4O5
InChI KeyKHPXUQMNIQBQEV-UHFFFAOYSA-N
SMILESOC(=O)CC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link