Oxalyl Monoguanylhydrazide 97%, Thermo Scientific Chemicals
Oxalyl Monoguanylhydrazide 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalyl Monoguanylhydrazide 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Catalog number 416631000
Price (USD)/ Each
108.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
108.70
Add to cart
Oxalyl Monoguanylhydrazide 97%, Thermo Scientific Chemicals
Catalog number416631000
Price (USD)/ Each
108.70
-
Add to cart
Specifications
Chemical Name or MaterialOxalyl monoguanylhydrazide
Name Note97%
CAS89797-67-1
ColorWhite
Infrared SpectrumAuthentic
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only