Thermo Scientific Chemicals

Palmitic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 416701000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Palmitic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 416701000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 416701000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
408-35-5
IUPAC Name
sodium hexadecanoate
Molecular Formula
C16H31NaO2
InChI Key
GGXKEBACDBNFAF-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCCCC([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %
Loss on drying
=<2 % (110°C, 4 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text