Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Paraformaldehyde, 96%, Thermo Scientific Chemicals

Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning
Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
25 g
500 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 416780030
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
3 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Catalog number416780030
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS30525-89-4
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or beads
Infrared spectrumConforms
Titration with HCl>=95 %
Ash=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only