Chemical Identifiers
CAS569-61-9
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC19H18ClN3
InChI KeyJUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1
View more