Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals
Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 416931000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS356-42-3
IUPAC Name2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
Molecular FormulaC6F10O3
InChI KeyXETRHNFRKCNWAJ-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only