1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%, Thermo Scientific Chemicals
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 416955000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
Add to cart
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%, Thermo Scientific Chemicals
Catalog number416955000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2-{[2-(dimethylamino)ethyl](methyl)amino}ethyl)dimethylamine
Molecular FormulaC9H23N3
InChI KeyUKODFQOELJFMII-UHFFFAOYSA-N
SMILESCN(C)CCN(C)CCN(C)C
Molecular Weight (g/mol)173.30
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4415 to 1.4435 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only