Thermo Scientific Chemicals

Petrolatum, white, pure, Thermo Scientific Chemicals

Catalog number: 417090010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Petrolatum, white, pure, Thermo Scientific Chemicals

Catalog number: 417090010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 417090010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8009-03-8
IUPAC Name
1,1,2-trimethyl-1H-benzo[e]indole
Molecular Formula
C15H15N
InChI Key
WJZSZXCWMATYFX-UHFFFAOYSA-N
SMILES
CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Appearance (Color)
White
Appearance (Form)
Paste
Infrared spectrum
Conforms
Solidification point
50°C to 56°C
Viscosity
5 to 8 mm²/s (at 100°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text