Phenolphthalein, For analysis ACS
Phenolphthalein, For analysis ACS
Thermo Scientific Chemicals

Phenolphthalein, For analysis ACS

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 417185000
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
Add to cart
Phenolphthalein, For analysis ACS
Catalog number417185000
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Clarity of alcohol solutionPasses test
Visual transition intervalFrom pH 8.0 (colorless) to pH 10 (red)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only