Thermo Scientific Chemicals

Phenol Red

Catalog number: 417240050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenol Red

Catalog number: 417240050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 417240050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-74-8
IUPAC Name
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H14O5S
InChI Key
BELBBZDIHDAJOR-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Visual transition interval
From pH 6.8 (yellow) to pH 8.2 (red)
Infrared spectrum
Conforms
Clarity of solution
Passes test
Appearance (Color)
Red
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text