Phenol Red sodium salt, ACS reagent
Phenol Red sodium salt, ACS reagent
Thermo Scientific Chemicals

Phenol Red sodium salt, ACS reagent

Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 417250100
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
Add to cart
Phenol Red sodium salt, ACS reagent
Catalog number417250100
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H13NaO5S
InChI KeyHKHYOKBQJILTEI-UHFFFAOYNA-M
SMILES[Na+].OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
Molecular Weight (g/mol)376.36
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to red-brown
Appearance (Form)Powder
Infrared spectrumConforms
Clarity of solutionPasses test
Visual transition intervalFrom pH 6.8 (yellow) to pH 8.2 (red)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only