Thermo Scientific Chemicals

4-Phenylazophenol, 97%, Thermo Scientific Chemicals

Catalog number: 417361000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Phenylazophenol, 97%, Thermo Scientific Chemicals

Catalog number: 417361000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 417361000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1689-82-3
IUPAC Name
4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Molecular Formula
C12H10N2O
InChI Key
JTSBGMZPPPULTA-UHFFFAOYSA-N
SMILES
O=C1C=CC(C=C1)=NNC1=CC=CC=C1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Color)
Brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text