Thermo Scientific Chemicals

p-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 417481000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

p-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 417481000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 417481000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-18-0
IUPAC Name
dihydrogen benzene-1,4-diamine dichloride
Molecular Formula
C6H10Cl2N2
InChI Key
IYXMNTLBLQNMLM-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C=C1
Appearance (Color)
White to yellow to purple-grey
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Argentometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text