Thermo Scientific Chemicals

Phenyl Ether-biphenyl Eutectic, Thermo Scientific Chemicals

Catalog number: 417520010
1 kg, Each
Thermo Scientific Chemicals

Phenyl Ether-biphenyl Eutectic, Thermo Scientific Chemicals

Catalog number: 417520010
1 kg, Each
Quantity
Catalog number: 417520010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8004-13-5
IUPAC Name
1,1'-biphenyl; phenoxybenzene
Molecular Formula
C24H20O
InChI Key
MHCVCKDNQYMGEX-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=CC=CC=C1.O(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.5885 to 1.5905 (20°C, 589 nm)
GC
mix of phenyl ether-biphenyl: approx 70%-30%

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text