Thermo Scientific Chemicals

(R)-1-Phenyl-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 417750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-1-Phenyl-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 417750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 417750050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
103548-16-9
IUPAC Name
(1R)-1-phenylpropane-1,3-diol
Molecular Formula
C9H12O2
InChI Key
RRVFYOSEKOTFOG-SECBINFHSA-N
SMILES
OCC[C@@H](O)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
62°C to 66°C
GC
>=97.5 %
Specific optical rotation
+53° to +56° (22°C, 589 nm)
Specific optical rotation
(c=1, 1,2-dichlorethane)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text