Thermo Scientific Chemicals

Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 417895000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 417895000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 417895000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51429-74-4
IUPAC Name
12-Molybdophosphoric acid
Molecular Formula
H3Mo12O40P
Molecular Weight (g/mol)
1825.40
Synonym
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
Appearance (Color)
Yellow to yellow-orange
Appearance (Form)
Adhering crystals
Heavy metals (as Pb)
=<50 ppm
Insoluble matter
=<0.01 %
Chloride (Cl)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text