Pinacol, 99%, Thermo Scientific Chemicals
Pinacol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pinacol, 99%, Thermo Scientific Chemicals

Pinacol, 99%, C6H14O2, CAS Number-76-09-5, pinacone, pinacol, unii-527qe7i5co, tetramethylethylene glycol, 2,3-dimethyl-2,3-butanediol, 2,3-dimethyl-2,3-dihydroxybutane, 1,1,2,2-tetramethylethylene glycol, 2,3-butanediol, 2,3-dimethyl, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-butane-2,3-diol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 418001000
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
Add to cart
Pinacol, 99%, Thermo Scientific Chemicals
Catalog number418001000
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dimethylbutane-2,3-diol
Molecular FormulaC6H14O2
InChI KeyIVDFJHOHABJVEH-UHFFFAOYSA-N
SMILESCC(C)(O)C(C)(C)O
Molecular Weight (g/mol)118.18
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Low melting crystalline solid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only