Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals
Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 418020250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS59997-51-2
IUPAC Name4,4-dimethyl-3-oxopentanenitrile
Molecular FormulaC7H11NO
InChI KeyMXZMACXOMZKYHJ-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)White to cream
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point66°C to 72°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only