Thermo Scientific Chemicals

Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 418025000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 418025000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 418025000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59997-51-2
IUPAC Name
4,4-dimethyl-3-oxopentanenitrile
Molecular Formula
C7H11NO
InChI Key
MXZMACXOMZKYHJ-UHFFFAOYSA-N
SMILES
CC(C)(C)C(=O)CC#N
GC
>=96.0 %
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
66°C to 72°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text