Poly(ethylene glycol), average M.W. 1450, Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 1450, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 1450, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 418040030
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
3 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
Add to cart
Poly(ethylene glycol), average M.W. 1450, Thermo Scientific Chemicals
Catalog number418040030
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Waxy flakes
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Hydroxyl value70 to 80 mg KOH/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only