Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals
Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
3 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 418050010
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals
Catalog number418050010
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Flakes
Water=<0.5 % (K.F.)
Hydroxyl value12 to 16 mg KOH/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only