Thermo Scientific Chemicals

Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals

Catalog number: 418050030
3 kg, Plastic drum, Each
Thermo Scientific Chemicals

Poly(ethylene Glycol), Average M.W. 8000, Thermo Scientific Chemicals

Catalog number: 418050030
3 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 418050030
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Flakes
Water
=<0.5 % (K.F.)
Hydroxyl value
12 to 16 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text