Acetic acid, potassium salt, 99+%, ACS reagent, Thermo Scientific Chemicals
Acetic acid, potassium salt, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 418170025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
Molecular Weight (g/mol)98.14
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Heavy metals (ICP-OES)=<5 ppm
Iron (Fe)=<5 ppm
Sodium (Na)=<0.03 %
Sulfate (SO4)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only