Thermo Scientific Chemicals

Potassium chloride, for biochemistry

Catalog number: 418205000
500 g, Each
Thermo Scientific Chemicals

Potassium chloride, for biochemistry

Catalog number: 418205000
500 g, Each
Quantity
Catalog number: 418205000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Appearance (Form)
Crystalline powder or crystals
Titration Argentometric
>=99.0 %
Solubility
(1 M in water) Clear colorless
Appearance (Color)
White
Magnesium (Mg)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for biochemistry
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text