Potassium formate, 96%, pract., Thermo Scientific Chemicals
Potassium formate, 96%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium formate, 96%, pract., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 418221000
Price (USD)/ Each
25.90
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
25.90
Add to cart
Potassium formate, 96%, pract., Thermo Scientific Chemicals
Catalog number418221000
Price (USD)/ Each
25.90
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium formate
Molecular FormulaCHKO2
InChI KeyWFIZEGIEIOHZCP-UHFFFAOYSA-M
SMILES[K+].[O-]C=O
Molecular Weight (g/mol)84.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=95.0 %
Melting point164°C to 168°C
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only