Potassium formate, 96%, pract.
Potassium formate, 96%, pract.
Potassium formate, 96%, pract.
Potassium formate, 96%, pract.
Thermo Scientific Chemicals

Potassium formate, 96%, pract.

CAS: 590-29-4 | CHKO2 | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 418225000
Price (USD)
59.65
Special offer
Online exclusive
Ends: 26-Sep-2025
73.20
Save 13.55 (19%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 26-Sep-2025
73.20
Save 13.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS590-29-4
IUPAC Namepotassium formate
Molecular FormulaCHKO2
InChI KeyWFIZEGIEIOHZCP-UHFFFAOYSA-M
SMILES[K+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=95.0 %
Melting point164°C to 168°C
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link