Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals
Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 418241000
Price (USD)/ Each
69.65
Online exclusive
77.00 
Save 7.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
77.00 
Save 7.35 (10%)
Add to cart
Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals
Catalog number418241000
Price (USD)/ Each
69.65
Online exclusive
77.00 
Save 7.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-05-6
IUPAC Namepotassium iodate
Molecular FormulaIKO3
InChI KeyJLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Chloride and Bromide=<0.01 % (as Cl)
Sulfate (SO4)=<0.005 %
Sodium (Na)=<0.005 %
pH5.0 to 8.0 (5% soln. at 25°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only