Thermo Scientific Chemicals

Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals

Catalog number: 418245000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals

Catalog number: 418245000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 418245000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-05-6
IUPAC Name
potassium iodate
Molecular Formula
IKO3
InChI Key
JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES
[K+].[O-][I](=O)=O
Appearance (Form)
Powder or crystalline powder
Chloride and Bromide
=<0.01 % (as Cl)
Sulfate (SO4)
=<0.005 %
Sodium (Na)
=<0.005 %
pH
5.0 to 8.0 (5% soln. at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text