Potassium iodide, 99+%, ACS reagent, Thermo Scientific Chemicals
Potassium iodide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium iodide, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Catalog number 418260050
Price (USD)/ Each
1,710.65
Online exclusive
1901.00 
Save 190.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
1,710.65
Online exclusive
1901.00 
Save 190.35 (10%)
Add to cart
Potassium iodide, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number418260050
Price (USD)/ Each
1,710.65
Online exclusive
1901.00 
Save 190.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals or granules
Titration with KIO3>=99.0 %
pH6.0 to 9.2 (5% soln. at 25°C)
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only