Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals
Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 418270025
Price (USD)/ Each
109.65
Online exclusive
129.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
129.00 
Save 19.35 (15%)
Add to cart
Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals
Catalog number418270025
Price (USD)/ Each
109.65
Online exclusive
129.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedipotassium sulfinatosulfonate
Molecular FormulaK2O5S2
InChI KeyRWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)S([O-])(=O)=O
Molecular Weight (g/mol)222.31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Titration Iodimetric>=96.0 %
Heavy metals (as Pb)=<10 ppm
Selenium (Se)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only