Thermo Scientific Chemicals

Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 418270250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 418270250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 418270250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16731-55-8
IUPAC Name
dipotassium sulfinatosulfonate
Molecular Formula
K2O5S2
InChI Key
RWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Titration Iodimetric
>=96.0 %
Heavy metals (as Pb)
=<10 ppm
Selenium (Se)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text