Potassium periodate, ACS reagent
Potassium periodate, ACS reagent
Potassium periodate, ACS reagent
Potassium periodate, ACS reagent
Thermo Scientific Chemicals

Potassium periodate, ACS reagent

CAS: 7790-21-8 | IKO4 | 230 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 418291000
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7790-21-8
IUPAC Namepotassium periodate
Molecular FormulaIKO4
InChI KeyFJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with Na2S2O399.8 to 100.3 % (dried basis)
Other halogens (as Cl)=<0.01 %
Manganese (Mn)=<1 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link