L(+)-Prolinol, 95%, Thermo Scientific Chemicals
L(+)-Prolinol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-Prolinol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 418320050
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.80 
Save 14.15 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.80 
Save 14.15 (15%)
Add to cart
L(+)-Prolinol, 95%, Thermo Scientific Chemicals
Catalog number418320050
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.80 
Save 14.15 (15%)
-
Add to cart
Chemical Identifiers
CAS23356-96-9
IUPAC Name(pyrrolidin-2-yl)methanol
Molecular FormulaC5H11NO
InChI KeyHVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILESOCC1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4840 to 1.4870 (20°C, 589 nm)
Specific optical rotation+29° to +33° (20°C, 589 nm) (c=1, toluene)
GC>=94 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only