Thermo Scientific Chemicals

1-Propanesulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 418340250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Propanesulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 418340250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 418340250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5284-66-2
IUPAC Name
propane-1-sulfonic acid
Molecular Formula
C3H8O3S
InChI Key
KCXFHTAICRTXLI-UHFFFAOYSA-N
SMILES
CCCS(O)(=O)=O
Appearance (Color)
Clear brown to dark brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration with NaOH
>=94 %
Refractive index
1.4380 to 1.4410 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text