Pulegone, 92%, pract.
Pulegone, 92%, pract.
Pulegone, 92%, pract.
Pulegone, 92%, pract.
Thermo Scientific Chemicals

Pulegone, 92%, pract.

Pulegone, CAS # 89-82-7, is a compound found in the oils of certain plant species including peppermint, catnip, and pennyroyal. | CAS: 89-82-7 | C10H16O | 152.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
418501000100 gGlass bottle
Catalog number 418501000
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS89-82-7
IUPAC Name(5R)-5-methyl-2-(propan-2-ylidene)cyclohexan-1-one
Molecular FormulaC10H16O
InChI KeyNZGWDASTMWDZIW-MRVPVSSYSA-N
SMILESC[C@@H]1CCC(=C(C)C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=90 %
Refractive index1.4850 to 1.4880 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations

Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Zhang, Z.; Xie, Y.; Wang ,Y.; Lin, Z.; Wang, L.; Li, G. Toxicities of monoterpenes against housefly, Musca domestica L. (Diptera: Muscidae). Environ Sci Pollut Res Int. 2017, 31, 24708-24713.
  2. Schomburg, D.; Schomburg, I. 2013, (+)-pulegone reductase 1.3.1.81. In: Schomburg, D.; Schomburg, I. (eds) Class 1 Oxidoreductases. Springer Handbook of Enzymes. Springer, Berlin, Heidelberg.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link