Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number 418540025
Price (USD)/ Each
339.65
Online exclusive
377.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
339.65
Online exclusive
377.00 
Save 37.35 (10%)
Add to cart
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number418540025
Price (USD)/ Each
339.65
Online exclusive
377.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
Molecular Weight (g/mol)79.10
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(in water) Passes test
Sulfate (SO4)=<10 ppm
Reducing substancesPasses test
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only