Thermo Scientific Chemicals

Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 418545000
500 mL, EcoSafPak™ Glass Bottle, Each
Thermo Scientific Chemicals

Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 418545000
500 mL, EcoSafPak™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 418545000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-86-1
IUPAC Name
pyridine
Molecular Formula
C5H5N
InChI Key
JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1
Appearance (Form)
Liquid
Water
=<0.1 % (K.F.)
Ammonia (NH3)
=<20 ppm
Appearance (Color)
Clear colorless
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text