Pyrogallol, ACS reagent, Thermo Scientific Chemicals
Pyrogallol, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrogallol, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 418621000
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
Pyrogallol, ACS reagent, Thermo Scientific Chemicals
Catalog number418621000
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS87-66-1
IUPAC Namebenzene-1,2,3-triol
Molecular FormulaC6H6O3
InChI KeyWQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Residue after ignition=<0.005 %
Sulfate (SO4)=<50 ppm
Appearance (Color)White
Melting point131.0°C to 135.0°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only