Pyronin Y, Certified
Pyronin Y, Certified
Thermo Scientific Chemicals

Pyronin Y, Certified

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 418630050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC17H19ClN2O
InChI KeyINCIMLINXXICKS-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC2=[O+]C3=CC(=CC=C3C=C2C=C1)N(C)C
Molecular Weight (g/mol)302.80
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Absorption Ratio P-15/P+151.23 to 1.7
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Dye content>=45 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only