Thermo Scientific Chemicals

2-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 418810100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 418810100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 418810100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
59-31-4
IUPAC Name
1,2-dihydroquinolin-2-one
Molecular Formula
C9H7NO
InChI Key
LISFMEBWQUVKPJ-UHFFFAOYSA-N
SMILES
O=C1NC2=CC=CC=C2C=C1
Appearance (Color)
White to brown-purple to purple
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Powder
Melting point
194°C to 200°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text