Thermo Scientific Chemicals

Rhodamine 6G 99%

Catalog number: 419010250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Rhodamine 6G 99%

Catalog number: 419010250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419010250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
989-38-8
IUPAC Name
hydrogen ethyl 2-[6-(ethylamino)-3-(ethylimino)-2,7-dimethyl-3H-xanthen-9-yl]benzoate chloride
Molecular Formula
C28H31ClN2O3
InChI Key
VYXSBFYARXAAKO-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCNC1=C(C)C=C2C(OC3=CC(=NCC)C(C)=CC3=C2C2=CC=CC=C2C(=O)OCC)=C1
Appearance (Color)
Red-brown to brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
UV spectrophotometry
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text