Chemical Identifiers
CAS13161-28-9
IUPAC Name9-[2-(ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethyl-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyXFKVYXCRNATCOO-UHFFFAOYSA-M
SMILES[Cl-].CCNC1=CC2=[O+]C3=CC(NCC)=C(C)C=C3C(C3=CC=CC=C3C(=O)OCC)=C2C=C1C
View more