Thermo Scientific Chemicals

Rhodamine 123, +99%

Catalog number: 419091000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

Rhodamine 123, +99%

Catalog number: 419091000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 419091000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62669-70-9
IUPAC Name
3,6-diamino-9-[2-(methoxycarbonyl)phenyl]-10λ⁴-xanthen-10-ylium chloride
Molecular Formula
C21H17ClN2O3
InChI Key
TUFFYSFVSYUHPA-UHFFFAOYSA-M
SMILES
[Cl-].COC(=O)C1=CC=CC=C1C1=C2C=CC(N)=CC2=[O+]C2=CC(N)=CC=C12
Appearance (Color)
Red-brown to brown
Infrared spectrum
Conforms
UV spectrophotometry
>=99.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text