Chemical Identifiers
CAS62669-70-9
IUPAC Name3,6-diamino-9-[2-(methoxycarbonyl)phenyl]-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC21H17ClN2O3
InChI KeyTUFFYSFVSYUHPA-UHFFFAOYSA-M
SMILES[Cl-].COC(=O)C1=CC=CC=C1C1=C2C=CC(N)=CC2=[O+]C2=CC(N)=CC=C12
View more