Thermo Scientific Chemicals

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh, Thermo Scientific Chemicals

Catalog number: 419100025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh, Thermo Scientific Chemicals

Catalog number: 419100025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419100025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10049-07-7
IUPAC Name
rhodium(3+) trichloride
Molecular Formula
Cl3Rh
InChI Key
SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Rh+3]
Appearance (Color)
Red, grey, red-black or black
Appearance (Form)
Crystalline powder, crystals or granules
Assay
48.6 to 49.2 % (Rh)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text