Thermo Scientific Chemicals

Basic Fuchsin, 88%, pure

Catalog number: 419140025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Basic Fuchsin, 88%, pure

Catalog number: 419140025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 419140025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
632-99-5
IUPAC Name
hydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular Formula
C20H20ClN3
InChI Key
AXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Appearance (Color)
Green
Appearance (Form)
Powder
Infrared spectrum
Conforms
Dye content
>=83 % (on dry substance)
UV Lambda max
549 to 555 nm (in 50:50 IMS:water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text