Chemical Identifiers
IUPAC Namedisodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular FormulaC20H2Cl4I4Na2O5
InChI KeyKCQREHTWEUECQT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
Molecular Weight (g/mol)1017.63
View more