Thermo Scientific Chemicals

Rosin, gum, Thermo Scientific Chemicals

Catalog number: 419170025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Rosin, gum, Thermo Scientific Chemicals

Catalog number: 419170025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 419170025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8050-09-7
IUPAC Name
1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular Formula
C20H30O2
InChI Key
RSWGJHLUYNHPMX-UHFFFAOYNA-N
SMILES
CC(C)C1=CC2=CCC3C(C)(CCCC3(C)C(O)=O)C2CC1
Appearance (Color)
Almost white to yellow to orange
Appearance (Form)
Powder or flakes or chunks
Infrared spectrum
Conforms
Acid value
156 to 182 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text